Methyl-1-benzofuran-2-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl benzofuran-2-carboxylate

OR303096-250MG
APOSOR303096-250MGEA 33.5 EUR
APOSOR303096-250MG
Methyl-1-benzofuran-2-carboxylate
Methyl-1-benzofuran-2-carboxylate
Formel: C₁₀H₈O₃
Molecular Weight: 176.17 g/mol
Schmelzpunkt: 53…55 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01159618
CAS-Nummer: 1646-27-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR