tert-Butyl-4-{[8-(ethoxycarbonyl)[1,2,4]triazolo[4,3-a]pyridin-3-yl]methyl}piperidine-1-carboxylate

Lieferant: Apollo Scientific

OR303120-500MG
APOSOR303120-500MGEA 661 EUR
APOSOR303120-500MG
tert-Butyl-4-{[8-(ethoxycarbonyl)[1,2,4]triazolo[4,3-a]pyridin-3-yl]methyl}piperidine-1-carboxylate
tert-Butyl-4-{[8-(ethoxycarbonyl)[1,2,4]triazolo[4,3-a]pyridin-3-yl]methyl}piperidine-1-carboxylate
Formel: C₂₀H₂₈N₄O₄
Molecular Weight: 388,47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12755391
CAS-Nummer: 1211592-94-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR