Methyl-6-amino-5-iodonicotinate
Lieferant: Apollo Scientific
Synonyme:
Methyl 6-amino-5-iodonicotinate, Methyl 6-amino-5-iodopyridine-3-carboxylate
OR303135-1G
OR303135-5G
APOSOR303135-1GEA
81.2
EUR
APOSOR303135-1G
APOSOR303135-5G
Methyl-6-amino-5-iodonicotinate
Methyl-6-amino-5-iodonicotinate
Formel:
C₇H₇IN₂O₂ Molecular Weight: 278.05 g/mol Schmelzpunkt: 164…167 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD16660021 CAS-Nummer: 211308-80-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...