Methyl 6-(4-methylpiperazin-1-yl)pyridine-3-carboxylate
Lieferant: Apollo Scientific
OR303142-1G
APOSOR303142-1GEA
0
EUR
APOSOR303142-1G
Methyl 6-(4-methylpiperazin-1-yl)pyridine-3-carboxylate
Methyl 6-(4-methylpiperazin-1-yl)pyridine-3-carboxylate
Formel:
C₁₂H₁₇N₃O₂ Molecular Weight: 235,282 g/mol Schmelzpunkt: 76…79 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00894965 CAS-Nummer: 132144-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...