5-Methoxy-1-benzothiophene-3-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR303166-1G OR303166-500MG
APOSOR303166-1GEA 671 EUR
APOSOR303166-1G APOSOR303166-500MG
5-Methoxy-1-benzothiophene-3-carboxylic acid 97%
5-Methoxy-1-benzothiophene-3-carboxylic acid
Formel: C₁₀H₈O₃S
Molecular Weight: 208.234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19440790
CAS-Nummer: 123392-43-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR