Methyl (3E)-4-(4-bromophenyl)-2-oxobut-3-enoate

Lieferant: Apollo Scientific

Synonyme: (E)-Methyl 4-(4-bromophenyl)-2-oxobut-3-enoate

OR303171-1G
APOSOR303171-1GEA 180 EUR
APOSOR303171-1G
Methyl (3E)-4-(4-bromophenyl)-2-oxobut-3-enoate
Methyl (3E)-4-(4-bromophenyl)-2-oxobut-3-enoate
Formel: C₁₁H₉BrO₃
Molecular Weight: 269,091 g/mol
Schmelzpunkt: 120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD18642374
CAS-Nummer: 608128-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR