4-Bromo-7-methoxy-2,2-dimethyl-2,3-dihydro-1-benzofuran
Lieferant: Apollo Scientific
|
Achtung
|
OR303176-1G
OR303176-500MG
APOSOR303176-1GEA
671
EUR
APOSOR303176-1G
APOSOR303176-500MG
4-Bromo-7-methoxy-2,2-dimethyl-2,3-dihydro-1-benzofuran
4-Bromo-7-methoxy-2,2-dimethyl-2,3-dihydro-1-benzofuran
Formel:
C₁₁H₁₃BrO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD22414513 CAS-Nummer: 185406-76-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...