(2E)-3-{1,4-Dioxaspiro[4.5]dec-7-en-7-yl}prop-2-enoic acid

Lieferant: Apollo Scientific

OR303181-500MG
APOSOR303181-500MGEA 661 EUR
APOSOR303181-500MG
(2E)-3-{1,4-Dioxaspiro[4.5]dec-7-en-7-yl}prop-2-enoic acid
(2E)-3-{1,4-Dioxaspiro[4.5]dec-7-en-7-yl}prop-2-enoic acid
Formel: C₁₁H₁₄O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD13152280
CAS-Nummer: 226916-19-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR