tert-Butyl 5-hydroxy-2-azabicyclo[2.2.1]heptane-2-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 5-hydroxy-2-aza-bicyclo[2.2.1]heptane-2-carboxylate

OR303189-100MG
APOSOR303189-100MGEA 21.9 EUR
APOSOR303189-100MG
tert-Butyl 5-hydroxy-2-azabicyclo[2.2.1]heptane-2-carboxylate
tert-Butyl 5-hydroxy-2-azabicyclo[2.2.1]heptane-2-carboxylate
Formel: C₁₁H₁₉NO₃
Molecular Weight: 213,273 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11865204
CAS-Nummer: 207405-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR