Allylamine, N-FMOC protected

Lieferant: Apollo Scientific
Achtung

OR303191-1G OR303191-500MG
APOSOR303191-1GEA 671 EUR
APOSOR303191-1G APOSOR303191-500MG
Allylamine, N-FMOC protected
Allylamine, N-FMOC protected
Formel: C₁₈H₁₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD14635842
CAS-Nummer: 856438-23-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR