18-(Benzyloxy)-18-oxooctadecanoic acid

Lieferant: Apollo Scientific
Achtung

OR303206-500MG
APOSOR303206-500MGEA 782 EUR
APOSOR303206-500MG
18-(Benzyloxy)-18-oxooctadecanoic acid
18-(Benzyloxy)-18-oxooctadecanoic acid
Formel: C₂₅H₄₀O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD09991736
CAS-Nummer: 189625-51-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR