tert-Butyl Hydrogen Hexadecanedioate 98%

Lieferant: Apollo Scientific
Achtung

OR303207-1G OR303207-5G OR303207-10G
APOSOR303207-1GEA 48 EUR
APOSOR303207-1G APOSOR303207-5G APOSOR303207-10G
tert-Butyl Hydrogen Hexadecanedioate 98%
tert-Butyl Hydrogen Hexadecanedioate
Formel: C₂₀H₃₈O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD23136039
CAS-Nummer: 843666-27-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR