tert-Butyl Hydrogen tetradecanedioate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
14-(tert-Butoxy)-14-oxotetradecanoic acid, Tetradecanedioic acid Mono-tert-butyl ester
OR303208-5G
APOSOR303208-5GEA
72.5
EUR
APOSOR303208-5G
tert-Butyl Hydrogen tetradecanedioate
tert-Butyl Hydrogen tetradecanedioate
Formel:
C₁₈H₃₄O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD23136040 CAS-Nummer: 234082-00-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...