5-Amino-3-methyl-1,2-thiazole-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR303238-1G OR303238-500MG
APOSOR303238-1GEA 336 EUR
APOSOR303238-1G APOSOR303238-500MG
5-Amino-3-methyl-1,2-thiazole-4-carboxylic acid
5-Amino-3-methyl-1,2-thiazole-4-carboxylic acid
Formel: C₅H₆N₂O₂S
Lagertemperatur: Kühlschrank
MDL: MFCD06376749
CAS-Nummer: 22131-51-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR