Methyl 2-(cyclohexylamino)-5-nitrobenzoate

Lieferant: Apollo Scientific

OR303242-1G OR303242-500MG
APOSOR303242-1GEA 180 EUR
APOSOR303242-1G APOSOR303242-500MG
Methyl 2-(cyclohexylamino)-5-nitrobenzoate
Methyl 2-(cyclohexylamino)-5-nitrobenzoate
Formel: C₁₄H₁₈N₂O₄
Molecular Weight: 278.304 g/mol
Schmelzpunkt: 132…134 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD23989316
CAS-Nummer: 83909-56-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR