4-Iod-1-phenyl-1H-pyrazol
Lieferant: Apollo Scientific
|
Achtung
|
OR303252-1G
OR303252-250MG
APOSOR303252-1GEA
190
EUR
APOSOR303252-1G
APOSOR303252-250MG
4-Iod-1-phenyl-1H-pyrazol
4-Iodo-1-phenyl-1H-pyrazole
Formel:
C₉H₇IN₂ Schmelzpunkt: 72…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11707168 CAS-Nummer: 23889-85-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...