3-cyclopropyl-1H-pyrazole 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Cyclopropyl-1H-pyrazole

OR303295-5G
APOSOR303295-5GEA 79.9 EUR
APOSOR303295-5G
3-cyclopropyl-1H-pyrazole 97+%
3-cyclopropyl-1H-pyrazole
Formel: C₆H₈N₂
Molecular Weight: 108.144 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD0497093
CAS-Nummer: 100114-57-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR