(1R)-5-tert-Butyl-2,3-dihydro-1H-inden-1-amine ≥ ee 99%

Lieferant: Apollo Scientific

OR303308-500MG
APOSOR303308-500MGEA 1350 EUR
APOSOR303308-500MG
(1R)-5-tert-Butyl-2,3-dihydro-1H-inden-1-amine ≥ ee 99%
(1R)-5-tert-Butyl-2,3-dihydro-1H-inden-1-amine
Formel: C₁₃H₁₉N
Molecular Weight: 189,297 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11515603
CAS-Nummer: 808756-83-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR