(1R,2R)-2-Methylcyclopropane-1-carboxylic acid 97+%

Lieferant: Apollo Scientific
Achtung

OR303309-500MG
APOSOR303309-500MGEA 560 EUR
APOSOR303309-500MG
(1R,2R)-2-Methylcyclopropane-1-carboxylic acid 97+%
(1R,2R)-2-Methylcyclopropane-1-carboxylic acid
Formel: C₅H₈O₂
Molecular Weight: 100,116 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19228131
CAS-Nummer: 10487-86-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR