Methyl-1H-indazol-3-carboxylat

Lieferant: Apollo Scientific
Achtung

OR303318-100G
APOSOR303318-100GEA 111 EUR
APOSOR303318-100G
Methyl-1H-indazol-3-carboxylat
Methyl-1H-indazol-3-carboxylat
Formel: C₉H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01138133
CAS-Nummer: 43120-28-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR