5-Bromo-1-methyl-1H-pyrazole-4-carboxylic acid ≥98%

Lieferant: Apollo Scientific

OR303319-1G
APOSOR303319-1GEA 104 EUR
APOSOR303319-1G
5-Bromo-1-methyl-1H-pyrazole-4-carboxylic acid ≥98%
5-Bromo-1-methyl-1H-pyrazole-4-carboxylic acid
Formel: C₅H₅BrN₂O₂
Molecular Weight: 205,009 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22052492
CAS-Nummer: 54367-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR