[(Trimethylsilyl)ethynyl]boronic acid, pinacol ester

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,4,5,5-Tetramethyl-2-[(trimethylsilyl)ethynyl]-1,3,2-dioxaborolane, Trimethyl((4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethynyl)silane, [(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)ethynyl]trimethylsilane

OR303320-250MG
APOSOR303320-250MGEA 72.5 EUR
APOSOR303320-250MG
[(Trimethylsilyl)ethynyl]boronic acid, pinacol ester
[(Trimethylsilyl)ethynyl]boronic acid, pinacol ester
Formel: C₁₁H₂₁BO₂Si
Molecular Weight: 224.18 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13182130
CAS-Nummer: 159087-46-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR