3-{2-[(4-Chlorophenyl)thio]-5-nitrophenyl}acrylic acid

Lieferant: Apollo Scientific

OR30333-10G OR30333-1G OR30333-5G
APOSOR30333-10GEA 519 EUR
APOSOR30333-10G APOSOR30333-1G APOSOR30333-5G
3-{2-[(4-Chlorophenyl)thio]-5-nitrophenyl}acrylic acid
3-{2-[(4-Chlorophenyl)thio]-5-nitrophenyl}acrylic acid
Formel: C₁₅H₁₀ClNO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD02090622
CAS-Nummer: 175278-51-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR