5-Chloro-1H-pyrazolo[3,4-b]pyridine

Lieferant: Apollo Scientific

OR303341-1G
APOSOR303341-1GEA 144 EUR
APOSOR303341-1G
5-Chloro-1H-pyrazolo[3,4-b]pyridine
5-Chloro-1H-pyrazolo[3,4-b]pyridine
Formel: C₆H₄ClN₃
Molecular Weight: 153.569 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11846318
CAS-Nummer: 1240725-66-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR