Ethyl potassium oxalate

Lieferant: Apollo Scientific
Achtung

OR303345-25G
APOSOR303345-25GEA 145 EUR
APOSOR303345-25G
Ethyl potassium oxalate
Ethyl potassium oxalate
Formel: C₄H₅KO₄
Schmelzpunkt: 220…223 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00045861
CAS-Nummer: 1906-57-6
EINECS: 217-606-8
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR