2-(Octadecyloxy)ethan-1-ol 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR303347-1G
OR303347-5G
APOSOR303347-1GEA
0
EUR
APOSOR303347-1G
APOSOR303347-5G
2-(Octadecyloxy)ethan-1-ol 95%
2-(Octadecyloxy)ethan-1-ol
Formel:
C₂₀H₄₂O₂ Schmelzpunkt: 49…52 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043351 CAS-Nummer: 2136-72-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...