2-Bromo-3-nonylthiophene

Lieferant: Apollo Scientific

OR303356-10G OR303356-1G OR303356-5G
APOSOR303356-10GEA 346 EUR
APOSOR303356-10G APOSOR303356-1G APOSOR303356-5G
2-Bromo-3-nonylthiophene
2-Bromo-3-nonylthiophene
Formel: C₁₃H₂₁BrS
Lagertemperatur: Raumtemperatur
MDL: MFCD20922797
CAS-Nummer: 1398044-31-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR