4-(Diethylcarbamoyl)phenylboronsäurepinakolester 98%
Lieferant: Apollo Scientific
Synonyme:
4-(N,N-Diethylaminocarbonyl)phenylboronic acid, pinacol ester, [4-(Diethylamine-1-carbonyl)phenyl]boronic acid pinacol ester, N,N-Diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
OR303437-100G
OR303437-25G
OR303437-5G
APOSOR303437-100GEA
644
EUR
APOSOR303437-100G
APOSOR303437-25G
APOSOR303437-5G
4-(Diethylcarbamoyl)phenylboronsäurepinakolester 98%
4-(Diethylcarbamoyl)phenylboronsäurepinakolester
Formel:
C₁₇H₂₆BNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06795673 CAS-Nummer: 325142-99-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...