4-(Diethylcarbamoyl)phenylboronsäurepinakolester 98%

Lieferant: Apollo Scientific

Synonyme: 4-(N,N-Diethylaminocarbonyl)phenylboronic acid, pinacol ester, [4-(Diethylamine-1-carbonyl)phenyl]boronic acid pinacol ester, N,N-Diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

OR303437-100G OR303437-25G OR303437-5G
APOSOR303437-100GEA 644 EUR
APOSOR303437-100G APOSOR303437-25G APOSOR303437-5G
4-(Diethylcarbamoyl)phenylboronsäurepinakolester 98%
4-(Diethylcarbamoyl)phenylboronsäurepinakolester
Formel: C₁₇H₂₆BNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06795673
CAS-Nummer: 325142-99-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR