4-(N,N-Diphenylamino)phenylboronsäurepinakolester

Lieferant: Apollo Scientific

Synonyme: 4-(Diphenylamino)phenylboronic acid Pinacol ester, 4-(Diphenylamino)benzeneboronic acid pinacol ester, Triphenylamine-4-boronic acid Pinacol ester, N,N-Diphenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 2-[4-(Diphenylamino)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR303494-5G
APOSOR303494-5GEA 68.3 EUR
APOSOR303494-5G
4-(N,N-Diphenylamino)phenylboronsäurepinakolester
4-(N,N-Diphenylamino)phenylboronsäurepinakolester
Formel: C₂₄H₂₆BNO₂
Molecular Weight: 371.29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13195770
CAS-Nummer: 267221-88-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR