9,9-Di-n-octylfluorene-2-boronic acid pinacol ester
Lieferant: Apollo Scientific
|
Achtung
|
OR303500-1G
APOSOR303500-1GEA
56.7
EUR
APOSOR303500-1G
9,9-Di-n-octylfluorene-2-boronic acid pinacol ester
9,9-Di-n-octylfluorene-2-boronic acid pinacol ester
Formel:
C₃₅H₅₃BO₂ Schmelzpunkt: 50…56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD16294553 CAS-Nummer: 302554-81-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...