9,9-Dimethylfluorene-2,7-diboronic acid bis(pinacol) ester

Lieferant: Apollo Scientific

OR303501-5G
APOSOR303501-5GEA 39.3 EUR
APOSOR303501-5G
9,9-Dimethylfluorene-2,7-diboronic acid bis(pinacol) ester
9,9-Dimethylfluorene-2,7-diboronic acid bis(pinacol) ester
Formel: C₂₇H₃₆B₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD08704230
CAS-Nummer: 325129-69-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR