9,9-Dimethylfluoren-2-boronic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (9,9-Dimethyl-9H-fluoren-2-yl)boronic acid

OR303506-25G
APOSOR303506-25GEA 75.4 EUR
APOSOR303506-25G
9,9-Dimethylfluoren-2-boronic acid 97%
9,9-Dimethylfluoren-2-boronic acid
Formel: C₁₅H₁₅BO₂
Molecular Weight: 238.09 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08704227
CAS-Nummer: 333432-28-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR