1,3,5-Tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,2',2''-(1,3,5-Benzenetriyl)tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolane), 1,3,5-Phenyltriboronic acid tris(pinacol) ester

OR303539-1G
APOSOR303539-1GEA 31.9 EUR
APOSOR303539-1G
1,3,5-Tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene 97%
1,3,5-Tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
Formel: C₂₄H₃₉B₃O₆
Molecular Weight: 456,00 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17167305
CAS-Nummer: 365564-05-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR