4-(1-Hydroxyethyl)benzeneboronic acid, pinacol ester 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanol, 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethan-1-ol
OR303570-1G
OR303570-500MG
APOSOR303570-1GEA
130
EUR
APOSOR303570-1G
APOSOR303570-500MG
4-(1-Hydroxyethyl)benzeneboronic acid, pinacol ester 95%
4-(1-Hydroxyethyl)benzeneboronic acid, pinacol ester
Formel:
C₁₄H₂₁BO₃ Schmelzpunkt: 79…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD18733422 CAS-Nummer: 1173922-30-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...