4-(1-Hydroxyethyl)benzeneboronic acid, pinacol ester 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanol, 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethan-1-ol

OR303570-1G OR303570-500MG
APOSOR303570-1GEA 130 EUR
APOSOR303570-1G APOSOR303570-500MG
4-(1-Hydroxyethyl)benzeneboronic acid, pinacol ester 95%
4-(1-Hydroxyethyl)benzeneboronic acid, pinacol ester
Formel: C₁₄H₂₁BO₃
Schmelzpunkt: 79…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD18733422
CAS-Nummer: 1173922-30-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR