2,5-Piperazindion 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Glycine anhydride, Piperazine-2,5-dione

OR30359-100G
APOSOR30359-100GEA 144 EUR
APOSOR30359-100G
2,5-Piperazindion 98%
2,5-Piperazindion
Formel: C₄H₆N₂O₂
Molecular Weight: 114.1 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006009
CAS-Nummer: 106-57-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR