2-[4-(3-Nitro-2-pyridinyl)piperazino]-1-ethanol

Lieferant: Apollo Scientific

OR303590-1G
APOSOR303590-1GEA 0 EUR
APOSOR303590-1G
2-[4-(3-Nitro-2-pyridinyl)piperazino]-1-ethanol
2-[4-(3-Nitro-2-pyridinyl)piperazino]-1-ethanol
Formel: C₁₁H₁₆N₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00278397
CAS-Nummer: 215434-62-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR