2-(Methylsulphonyl)-3-nitropyridine
Lieferant: Apollo Scientific
OR303635-500MG
APOSOR303635-500MGEA
171
EUR
APOSOR303635-500MG
2-(Methylsulphonyl)-3-nitropyridine
2-(Methylsulphonyl)-3-nitropyridine
Formel:
C₆H₆N₂O₄S Molecular Weight: 202,19 g/mol Schmelzpunkt: 107…109 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD16140367 CAS-Nummer: 56825-29-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...