Methyl-5-chloro-6-methoxynicotinate
Lieferant: Apollo Scientific
OR303664-1G
APOSOR303664-1GEA
0
EUR
APOSOR303664-1G
Methyl-5-chloro-6-methoxynicotinate
Methyl-5-chloro-6-methoxynicotinate
Formel:
C₈H₈ClNO₃ Schmelzpunkt: 108…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12025914 CAS-Nummer: 220656-93-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...