5-(5-Chloro-2-pyridinyl)-1H-1,2,4-triazol-3-amine

Lieferant: Apollo Scientific
Achtung

OR303666-500MG
APOSOR303666-500MGEA 69.6 EUR
APOSOR303666-500MG
5-(5-Chloro-2-pyridinyl)-1H-1,2,4-triazol-3-amine
5-(5-Chloro-2-pyridinyl)-1H-1,2,4-triazol-3-amine
Formel: C₇H₆ClN₅
Molecular Weight: 195,61 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11100211
CAS-Nummer: 168893-69-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR