Methyl-5-chlorpicolinat 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 5-chloropyridine-2-carboxylate
OR303670-5G
APOSOR303670-5GEA
21.9
EUR
APOSOR303670-5G
Methyl-5-chlorpicolinat 97%
Methyl-5-chlorpicolinat
Formel:
C₇H₆ClNO₂ Molecular Weight: 171,5800 g/mol Schmelzpunkt: 80…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09702466 CAS-Nummer: 132308-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...