2-Chloro-N'-hydroxy-4-pyridinecarboximidamide
Lieferant: Apollo Scientific
OR303690-1G
APOSOR303690-1GEA
0
EUR
APOSOR303690-1G
2-Chloro-N'-hydroxy-4-pyridinecarboximidamide
2-Chloro-N'-hydroxy-4-pyridinecarboximidamide
Formel:
C₆H₆ClN₃O Schmelzpunkt: 162…164 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11052368 CAS-Nummer: 857653-85-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...