4,6-Dichloronicotinonitrile 98%

Lieferant: Apollo Scientific
Achtung

OR303715-25G
APOSOR303715-25GEA 87 EUR
APOSOR303715-25G
4,6-Dichloronicotinonitrile 98%
4,6-Dichloronicotinonitrile
Formel: C₆H₂Cl₂N₂
Molecular Weight: 173 g/mol
Schmelzpunkt: 133…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08276124
CAS-Nummer: 166526-03-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR