2-sulphanylnicotinonitrile

Lieferant: Apollo Scientific

OR303718-1G
APOSOR303718-1GEA 108 EUR
APOSOR303718-1G
2-sulphanylnicotinonitrile
2-sulphanylnicotinonitrile
Formel: C₆H₄N₂S
Molecular Weight: 136.18 g/mol
Schmelzpunkt: 248…250 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07343855
CAS-Nummer: 52505-45-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR