Methyl-4,6-dichloro-5-nitronicotinate 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR303727-1G
APOSOR303727-1GEA
538
EUR
APOSOR303727-1G
Methyl-4,6-dichloro-5-nitronicotinate 95%
Methyl-4,6-dichloro-5-nitronicotinate
Formel:
C₇H₄Cl₂N₂O₄ Schmelzpunkt: 65…66 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD13193110 CAS-Nummer: 89247-05-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...