2-Chloro-4-methyl-6,7-dihydro-5H-cyclopenta[b]pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-4-methyl-5H,6H,7H-cyclopenta[b]pyridine

OR303736-1G
APOSOR303736-1GEA 538 EUR
APOSOR303736-1G
2-Chloro-4-methyl-6,7-dihydro-5H-cyclopenta[b]pyridine
2-Chloro-4-methyl-6,7-dihydro-5H-cyclopenta[b]pyridine
Formel: C₉H₁₀ClN
Molecular Weight: 167,64 g/mol
Schmelzpunkt: 47…49 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08056605
CAS-Nummer: 267242-99-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR