6-(Phenylsulphanyl)-3-pyridinamine

Lieferant: Apollo Scientific

OR303738-500MG
APOSOR303738-500MGEA 336 EUR
APOSOR303738-500MG
6-(Phenylsulphanyl)-3-pyridinamine
6-(Phenylsulphanyl)-3-pyridinamine
Formel: C₁₁H₁₀N₂S
Molecular Weight: 202.28 g/mol
Schmelzpunkt: 118…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03230019
CAS-Nummer: 103983-07-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR