5-Chloro-2-pyridinecarbohydrazide 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloropicolinohydrazide

OR303749-1G
APOSOR303749-1GEA 342 EUR
APOSOR303749-1G
5-Chloro-2-pyridinecarbohydrazide 95%
5-Chloro-2-pyridinecarbohydrazide
Formel: C₆H₆ClN₃O
Molecular Weight: 171,59 g/mol
Schmelzpunkt: 178…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16622817
CAS-Nummer: 145835-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR