(6-Bromo-5-methoxy-2-pyridinyl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(6-Bromo-5-methoxypyridin-2-yl)methanol
OR303764-250MG
APOSOR303764-250MGEA
331
EUR
APOSOR303764-250MG
(6-Bromo-5-methoxy-2-pyridinyl)methanol
(6-Bromo-5-methoxy-2-pyridinyl)methanol
Formel:
C₇H₈BrNO₂ Molecular Weight: 218,06 g/mol Schmelzpunkt: 75…78 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09702159 CAS-Nummer: 905562-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...